1,2-Dibromo-4-fluorobenzene ≥98.0% (通过GC)
供应商: TCI
|
Warning
|
D2854-25G
D2854-5G
TCIAD2854-25GEA
0
CNY
TCIAD2854-25G
TCIAD2854-5G
1,2-Dibromo-4-fluorobenzene ≥98.0% (通过GC)
1,2-Dibromo-4-fluorobenzene
分子式:
C₆H₃Br₂F |
MDL编号:
MFCD00070780 CAS号: 2369-37-1 EINECS: 219-131-1 |
|
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...