4-溴-2-氯苯胺 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
B1993-25G
B1993-5G
TCIAB1993-25GEA
0
CNY
TCIAB1993-25G
TCIAB1993-5G
4-溴-2-氯苯胺 ≥98.0% (by GC)
4-溴-2-氯苯胺
分子式:
C₆H₅BrClN 分子量: 206.47 g/mol 沸点: 264 溶点: 71 存储条件: Ambient |
MDL编号:
MFCD00007660 CAS号: 38762-41-3 EINECS: 254-118-4 |
|
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 69.0 to 72.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...