2-溴-4-氯苯胺 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
B2268-25G
B2268-5G
TCIAB2268-25GEA
0
CNY
TCIAB2268-25G
TCIAB2268-5G
2-溴-4-氯苯胺 ≥98.0% (by GC)
2-溴-4-氯苯胺
分子式:
C₆H₅BrClN 分子量: 206.47 g/mol 沸点: 127°C 溶点: 69 |
MDL编号:
MFCD00041313 CAS号: 873-38-1 EINECS: 212-837-0 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 64.0 to 68.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...