1-溴-2,6-二氯苯 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 2-Bromo-1,3-dichlorobenzene

B0982-25G B0982-5G
TCIAB0982-25GEA 0 CNY
TCIAB0982-25G TCIAB0982-5G
1-溴-2,6-二氯苯 ≥98.0% (by GC)
1-溴-2,6-二氯苯
分子式: C₆H₃BrCl₂
分子量: 225.9 g/mol
沸点: 242°C
溶点: 66
MDL编号: MFCD00000574
CAS号: 19393-92-1
EINECS: 243-018-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a5d92c20301410a54bbd268fea2070723441857d&vid=75049dbcc1a12e2edccb47a217fc44079438ffaf&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 65.0 to 67.0 deg-C
Solubility in Methanol within almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR