2-氨基-5-溴苯甲酸 ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Warning

Synonyms: 5-溴氨茴酸

A1254-25G A1254-5G
TCIAA1254-25GEA 0 CNY
TCIAA1254-25G TCIAA1254-5G
2-氨基-5-溴苯甲酸 ≥98.0% (by HPLC, titration analysis)
2-氨基-5-溴苯甲酸
分子式: C₇H₆BrNO₂
分子量: 216.03 g/mol
沸点: 342
存储条件: Ambient
MDL编号: MFCD00007823
CAS号: 5794-88-7
EINECS: 227-338-3
Merck Index: 14,01405
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0a84de301cf60017511bacfc12aa9a442adff38&vid=e725dab4d1e3af0405b62d7387facf28d6c5bf49&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light red to Green powder to crystalline
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR