6-溴藜芦醛 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 6-Bromo-3,4-dimethoxybenzaldehyde, 2-Bromo-4,5-dimethoxybenzaldehyde

B2011-25G B2011-5G
TCIAB2011-25GEA 0 CNY
TCIAB2011-25G TCIAB2011-5G
6-溴藜芦醛 ≥98.0% (by GC)
6-溴藜芦醛
分子式: C₉H₉BrO₃
分子量: 245.07 g/mol
沸点: 300
溶点: 151
MDL编号: MFCD00003301
CAS号: 5392-10-9
EINECS: 226-390-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e7d466b8299b1f914b9ed49fe7e95183891d4f0&vid=b52d7b0e9092768190f0d027d618749253ac0a55&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 146.0 to 152.0 deg-C
Solubility in Toluene very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR