3-溴-4-甲氧基苯甲醛 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 3-溴-对茴香醛

B1790-25G B1790-5G
TCIAB1790-25GEA 0 CNY
TCIAB1790-25G TCIAB1790-5G
3-溴-4-甲氧基苯甲醛 ≥98.0% (by GC)
3-溴-4-甲氧基苯甲醛
分子式: C₈H₇BrO₂
分子量: 215.05 g/mol
沸点: 162°C
溶点: 54
MDL编号: MFCD00016599
CAS号: 34841-06-0
EINECS: 252-241-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a97862195eeff4a8ee4bbee455178619f3eeda7&vid=f7b3c8edc1b2418b434333131457034e5576e4b7&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 50.0 to 54.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR