4-氯苯基甲基砜 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
4-Chlorophenyl Methyl sulphone, 1-Chloro-4-(methylsulphonyl)benzene
C1237-25G
C1237-5G
TCIAC1237-25GEA
0
CNY
TCIAC1237-25G
TCIAC1237-5G
4-氯苯基甲基砜 ≥98.0% (by GC)
4-氯苯基甲基砜
分子式:
C₇H₇ClO₂S 分子量: 190.65 g/mol 沸点: 60°C |
MDL编号:
MFCD00025067 CAS号: 98-57-7 EINECS: 202-682-7 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Melting point | 98.0 to 101.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...