4-溴苯甲砜 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
1-Bromo-4-(methylsulphonyl)benzene
B3148-25G
B3148-5G
TCIAB3148-25GEA
0
CNY
TCIAB3148-25G
TCIAB3148-5G
4-溴苯甲砜 ≥98.0% (by GC)
4-溴苯甲砜
分子式:
C₇H₇BrO₂S 分子量: 235.1 g/mol |
MDL编号:
MFCD00025065 CAS号: 3466-32-8 EINECS: 222-421-0 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 102.0 to 106.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...