Methyl triacontanate ≥95.0% (by GC)
供应商: TCI
Synonyms:
Mellisic acid methyl ester, Triacontanoic acid methyl ester
T0812-1G
T0812-100MG
TCIAT0812-1GEA
0
CNY
TCIAT0812-1G
TCIAT0812-100MG
Methyl triacontanate ≥95.0% (by GC)
Methyl triacontanate
分子式:
C₃₁H₆₂O₂ 分子量: 466.83 g/mol |
MDL编号:
MFCD00042897 CAS号: 629-83-4 EINECS: 211-113-1 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 70.0 to 74.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...