2-丁基-2-乙基-1,3-丙二醇 ≥98.0% (by GC)
供应商: TCI
Synonyms:
2-Ethyl-2-(hydroxymethyl)-1-hexanol, 3,3-Bis(hydroxymethyl)heptane, 2-Butyl-2-ethylpropane-1,3-diol
B0922-25G
B0922-500G
TCIAB0922-25GEA
0
CNY
TCIAB0922-25G
TCIAB0922-500G
2-丁基-2-乙基-1,3-丙二醇 ≥98.0% (by GC)
2-丁基-2-乙基-1,3-丙二醇
分子式:
C₉H₂₀O₂ 沸点: 262 °C (1013 hPa) 溶点: 39 °C 密度: 0,931 g/cm³ (50 °C) |
MDL编号:
MFCD00004697 CAS号: 115-84-4 EINECS: 204-111-7 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 41.0 to 45.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...