9-蒽甲酸 ≥97.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
9-蒽羧酸
A0690-25G
A0690-5G
TCIAA0690-25GEA
0
CNY
TCIAA0690-25G
TCIAA0690-5G
9-蒽甲酸 ≥97.0% (by GC, titration analysis)
9-蒽甲酸
分子式:
C₁₅H₁₀O₂ 分子量: 222.24 g/mol 沸点: 411 溶点: 217°C 存储条件: Ambient |
MDL编号:
MFCD00001257 CAS号: 723-62-6 EINECS: 211-964-9 |
|
产品技术规格
Appearance | Light yellow to Yellow to Green powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...