9-溴蒽 ≥95.0% (by GC)

供应商: TCI
Warning

B0872-25G B0872-5G
TCIAB0872-25GEA 0 CNY
TCIAB0872-25G TCIAB0872-5G
9-溴蒽 ≥95.0% (by GC)
9-溴蒽
分子式: C₁₄H₉Br
分子量: 257.13 g/mol
沸点: 359
溶点: 103
存储条件: Ambient
MDL编号: MFCD00001243
CAS号: 1564-64-3
EINECS: 216-359-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a566956ba9da6b8ab581ecaedfcd86e9937c5b6f&vid=864cc8402a971508e9a7a4935630aca7f08be299&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 95.0 %
Melting point 99.0 to 104.0 deg-C
Solubility in Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR