1-十八烯 ≥90.0% (by GC)
供应商: TCI
Danger
O0008-25ML O0008-500ML
TCIAO0008-25MLEA 0 CNY
TCIAO0008-25ML TCIAO0008-500ML
1-十八烯 ≥90.0% (by GC)
1-十八烯
分子式: C₁₈H₃₆
分子量: 252.48 g/mol
沸点: 179°C
溶点: 17.5
闪点: 149°C
存储条件: Ambient
MDL编号: MFCD00009003
CAS号: 112-88-9
EINECS: 204-012-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a3a87e0ba6a21ea57192a0850b3ece30977fffc&vid=72dafb8b127f1ca3601d639b1a7574b0ccf91db5&prid=web&clid=SAPDAM
现在订购
产品技术规格
Appearance White to Almost white powder to lump
Purity (GC) min. 90.0 %