5-甲基-3-庚酮 ≥95.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
乙基异戊基酮
M0335-25ML
M0335-500ML
TCIAM0335-25MLEA
0
CNY
TCIAM0335-25ML
TCIAM0335-500ML
5-甲基-3-庚酮 ≥95.0% (by GC)
5-甲基-3-庚酮
分子式:
C₈H₁₆O 沸点: 157°C 密度: 0,82 g/cm³ (20 °C) 闪点: 59°C 存储条件: Ambient |
MDL编号:
MFCD00009340 CAS号: 541-85-5 EINECS: 208-793-7 UN号: 2271 ADR: 3,III Merck Index: 14,06084 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...