5-Methyl-2-octanone ≥80.0% (通过GC)
供应商: TCI
|
Warning
|
M0278-25ML
M0278-500ML
TCIAM0278-25MLEA
0
CNY
TCIAM0278-25ML
TCIAM0278-500ML
5-Methyl-2-octanone ≥80.0% (通过GC)
5-Methyl-2-octanone
分子式:
C₉H₁₈O 沸点: 102°C |
MDL编号:
MFCD00059414 CAS号: 58654-67-4 EINECS: 261-382-4 UN号: 1224 ADR: 3,III |
|
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 80.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...