1-癸硫醇 ≥95.0% (by GC, titration analysis)
供应商: TCI
|
Danger
|
Synonyms:
正癸硫醇, DT, 癸硫醇
D0016-25ML
D0016-500ML
TCIAD0016-25MLEA
0
CNY
TCIAD0016-25ML
TCIAD0016-500ML
1-癸硫醇 ≥95.0% (by GC, titration analysis)
1-癸硫醇
分子式:
C₁₀H₂₂S 分子量: 174.35 g/mol 沸点: 241°C 溶点: –26°C 闪点: 98°C 存储条件: Refrigerator |
MDL编号:
MFCD00004884 CAS号: 143-10-2 EINECS: 205-584-2 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Purity (Neutralization titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...