1,10-癸二醇 ≥97.0% (by GC)
供应商: TCI
Synonyms:
十次甲基二醇
D0014-25G
D0014-500G
TCIAD0014-25GEA
0
CNY
TCIAD0014-25G
TCIAD0014-500G
1,10-癸二醇 ≥97.0% (by GC)
1,10-癸二醇
分子式:
C₁₀H₂₂O₂ 分子量: 174.28 g/mol 沸点: 297 溶点: 72 存储条件: Ambient |
MDL编号:
MFCD00004749 CAS号: 112-47-0 EINECS: 203-975-2 Merck Index: 14,02849 |
|
产品技术规格
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 97.0 % |
Melting point | 72.0 to 75.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...