1-戊醇 ≥99.0% (by GC)

供应商: TCI
Warning

Synonyms: 戊醇, 1-戊醇, C5醇, 正戊醇

P0055-25ML P0055-500ML
TCIAP0055-25MLEA 0 CNY
TCIAP0055-25ML TCIAP0055-500ML
1-戊醇 ≥99.0% (by GC)
1-戊醇
分子式: H₃C(CH₂)₃CH₂OH
分子量: 88.15 g/mol
沸点: 137°C
溶点: –79°C
闪点: 43°C
存储条件: Ambient
MDL编号: MFCD00002977
CAS号: 71-41-0
EINECS: 200-752-1
UN号: 1105
ADR: 3 III
Merck Index: 14,07118
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14bb2c139339e85f43ccf9c77bced226ece2caaa&vid=a4bfbda0c57603519c4d829b37947a303c3ae6a2&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR