1-十六烷醇 ≥98.0% (通过GC)
供应商: TCI
H0071-500G
H0071-25G
TCIAH0071-500GEA
0
CNY
TCIAH0071-500G
TCIAH0071-25G
1-十六烷醇 ≥98.0% (通过GC)
1-十六烷醇
分子式:
H₃C(CH₂)₁₄CH₂OH 沸点: 305 - 330 °C (1013 hPa) 溶点: 46…52 °C 密度: 0,81 g/cm³ (20 °C) 闪点: ≥ 110 °C 存储条件: Ambient |
MDL编号:
MFCD00004760 CAS号: 36653-82-4 EINECS: 253-149-0 Merck Index: 14,02028 |
|
产品技术规格
Appearance | White powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 49.0 to 51.5 deg-C |
Solubility in EtOH | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...