正庚醇 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 庚醇

H0033-25ML H0033-500ML
TCIAH0033-25MLEA 0 CNY
TCIAH0033-25ML TCIAH0033-500ML
正庚醇 ≥98.0% (by GC)
正庚醇
分子式: H₃C(CH₂)₅CH₂OH
分子量: 116.2 g/mol
沸点: 177°C
溶点: –34°C
闪点: 71°C
存储条件: Ambient
MDL编号: MFCD00002986
CAS号: 111-70-6
EINECS: 203-897-9
Merck Index: 14,04661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d94fb43809b000bdd5da7153a0758c73437d766&vid=17a1cce2592226e10b4f96f56b5eabbd912d2745&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR