2,2-二甲氧基丙烷 ≥98.0% (通过GC)

供应商: TCI
Danger

A0057-25ML A0057-500ML
TCIAA0057-25MLEA 0 CNY
TCIAA0057-25ML TCIAA0057-500ML
2,2-二甲氧基丙烷 ≥98.0% (通过GC)
2,2-二甲氧基丙烷
分子式: C₅H₁₂O₂
沸点: 80°C
溶点: –47 °C
密度: 0,845 g/cm³ (20 °C)
闪点: -9°C
存储条件: Ambient
MDL编号: MFCD00008479
CAS号: 77-76-9
EINECS: 201-056-0
UN号: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6936f6b50ad13ab1937b8330817793655823a65&vid=9b0a772e59fcf58b8b5469cf364cbb732b4c4bc8&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR