十三烷胺 ≥98.0% (by GC, titration analysis)

供应商: TCI
Danger

Synonyms: 正十三胺

A0762-10G A0762-25G
TCIAA0762-10GEA 0 CNY
TCIAA0762-10G TCIAA0762-25G
十三烷胺 ≥98.0% (by GC, titration analysis)
十三烷胺
分子式: H₃C(CH₂)₁₁CH₂NH₂
分子量: 199.38 g/mol
沸点: 275°C
溶点: 28.1
存储条件: Ambient
MDL编号: MFCD00008156
CAS号: 2869-34-3
EINECS: 220-694-0
UN号: 3259
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64130a9f9b97dc4fcd9004fbfffdb2470a7f23a6&vid=d5dab8655a95a81e8c91c06167b29df637573166&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR