氯甲基乙醚 ≥96.0% (by GC)

供应商: TCI
Danger

Synonyms: Chloromethyl ethyl ether

C0201-25G C0201-500G
TCIAC0201-25GEA 0 CNY
TCIAC0201-25G TCIAC0201-500G
氯甲基乙醚 ≥96.0% (by GC)
氯甲基乙醚
分子式: ClCH₂OCH₂CH₃
沸点: 80°C
溶点: –98 °C
密度: 1,0188 g/cm³ (15 °C)
闪点: 7°C
存储条件: Refrigerator
MDL编号: MFCD00000887
CAS号: 3188-13-4
EINECS: 221-687-5
UN号: 2354
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5cb737e98551ae005790ae1c446287f4ead64b4b&vid=f8883037e1ae6b03836ed88027f50b15fc0700cd&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR