氯代十八烷 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
十八烷基氯, 硬脂基氯
C0235-500ML
C0235-25ML
TCIAC0235-500MLEA
0
CNY
TCIAC0235-500ML
TCIAC0235-25ML
氯代十八烷 ≥98.0% (by GC)
氯代十八烷
分子式:
C₁₈H₃₇Cl 分子量: 288.94 g/mol 沸点: 356 闪点: 184°C 存储条件: Refrigerator |
MDL编号:
MFCD00000960 CAS号: 3386-33-2 EINECS: 222-207-7 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Solubility in Toluene | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...