1-氯壬烷 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 壬基氯

C0233-25ML C0233-500ML
TCIAC0233-25MLEA 0 CNY
TCIAC0233-25ML TCIAC0233-500ML
1-氯壬烷 ≥98.0% (by GC)
1-氯壬烷
分子式: C₉H₁₉Cl
沸点: 204°C
密度: 0,87 g/cm³ (20 °C)
闪点: 74°C
存储条件: Ambient
MDL编号: MFCD00001023
CAS号: 2473-01-0
EINECS: 219-595-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48cd720eb53f004720212c25d875fb99fce2268c&vid=d828c0d85e43425a387b6d7809265448151ca28e&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR