1-氯十六烷 ≥98.0% (by GC)

供应商: TCI

Synonyms: 鲸蜡基氯, 十六烷基氯

C0183-25ML C0183-500ML
TCIAC0183-25MLEA 0 CNY
TCIAC0183-25ML TCIAC0183-500ML
1-氯十六烷 ≥98.0% (by GC)
1-氯十六烷
分子式: C₁₆H₃₃Cl
分子量: 260.89 g/mol
沸点: 195°C
溶点: 7.8
闪点: 136°C
存储条件: Ambient
MDL编号: MFCD00000959
CAS号: 4860-03-1
EINECS: 225-461-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b7be112e16421dadac05366b60090a512c6dd57&vid=f12c1ebd78e595cab705ce2f128c4437cc5521fb&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR