1,10-二氯癸烷 ≥97.0% (by GC)

供应商: TCI
Warning

Synonyms: 1,10-Decamethylene dichloride, Decane, 1,10-dichloro-

D0247-25G D0247-500G
TCIAD0247-25GEA 0 CNY
TCIAD0247-25G TCIAD0247-500G
1,10-二氯癸烷 ≥97.0% (by GC)
1,10-二氯癸烷
分子式: ClH₂C(CH₂)₈CH₂Cl
分子量: 211.17 g/mol
沸点: 102°C
闪点: 146°C
存储条件: Ambient
MDL编号: MFCD00000957
CAS号: 2162-98-3
EINECS: 218-489-6
UN号: 3082
ADR: 9 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0505841c9f24fd3381adfa0217b27ef58ef20086&vid=cf198f0ee30ad653043871965088e28aeb38c56d&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR