1,1,2-三氯乙烷 ≥98.0% (by GC)
供应商: TCI
Warning
T0381-25G T0381-500G
TCIAT0381-25GEA 0 CNY
TCIAT0381-25G TCIAT0381-500G
1,1,2-三氯乙烷 ≥98.0% (by GC)
1,1,2-三氯乙烷
分子式: Cl₂HCCH₂Cl
分子量: 133.4 g/mol
沸点: 114°C
溶点: -35.5
存储条件: Ambient
MDL编号: MFCD00000852
CAS号: 79-00-5
EINECS: 201-166-9
UN号: 2810
ADR: 6.1 III
Merck Index: 14,09637
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e79cb7c337faa9c9df90e95feaf86dfe5418624e&vid=d9a4e1651d694bdb99d47bf6a25919d6ccd73db7&prid=web&clid=SAPDAM
现在订购
产品技术规格
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %