十五烷酸 ≥98.0% (通过GC,滴定分析)

供应商: TCI
Warning

P0035-25G P0035-500G
TCIAP0035-25GEA 0 CNY
TCIAP0035-25G TCIAP0035-500G
十五烷酸 ≥98.0% (通过GC,滴定分析)
十五烷酸
分子式: C₁₅H₃₀O₂
沸点: 339°C
溶点: 51…53 °C
密度: 0,8496 g/cm³ (70 °C)
存储条件: Ambient
MDL编号: MFCD00002745
CAS号: 1002-84-2
EINECS: 213-693-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f0be94d8c97f2ad6e68556e7e3257af7858e23dc&vid=efa6575961f3ede48df6c7ba63429c8abd96aa12&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystaline
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 51.0 to 54.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR