正辛酸 ≥98.0% (by GC, titration analysis)

供应商: TCI
Danger

Synonyms: 羊脂酸, 辛酸

O0027-25ML O0027-500ML
TCIAO0027-25MLEA 0 CNY
TCIAO0027-25ML TCIAO0027-500ML
正辛酸 ≥98.0% (by GC, titration analysis)
正辛酸
分子式: C₈H₁₆O₂
沸点: 238°C
溶点: 16 °C
密度: 0,9066 g/cm³ (20 °C)
闪点: 109°C
存储条件: Ambient
MDL编号: MFCD00004429
CAS号: 124-07-2
EINECS: 204-677-5
UN号: 3265
ADR: 8,III
Merck Index: 14,01765
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8baff9f55c6508039e8ff1f7c22673d6be99ea8c&vid=2edb695e0219190de2a1295502164a676b16d239&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR