庚酸 ≥98.0% (by titrimetric analysis)
供应商: TCI
|
Danger
|
Synonyms:
葡萄花酸, 正庚酸
H0030-25ML
H0030-500ML
TCIAH0030-25MLEA
0
CNY
TCIAH0030-25ML
TCIAH0030-500ML
庚酸 ≥98.0% (by titrimetric analysis)
庚酸
分子式:
C₇H₁₄O₂ 分子量: 130.19 g/mol 沸点: 223°C 溶点: –8°C 闪点: 120°C 存储条件: Ambient |
MDL编号:
MFCD00004426 CAS号: 111-14-8 EINECS: 203-838-7 UN号: 3265 ADR: 8 II Merck Index: 14,04660 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 96.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...