Heptacosanoic acid ≥98.0% (通过滴定分析)

供应商: TCI

H0971-1G H0971-100MG H0971-5G
TCIAH0971-1GEA 0 CNY
TCIAH0971-1G TCIAH0971-100MG TCIAH0971-5G
Heptacosanoic acid ≥98.0% (通过滴定分析)
Heptacosanoic acid
分子式: C₂₇H₅₄O₂
存储条件: Ambient
MDL编号: MFCD00014029
CAS号: 7138-40-1
EINECS: 230-436-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48fafcfb56995b05b0166ca07b72d74aebe6130f&vid=a2946d4001135127a2534b9a8492ebea37f64163&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 90.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 84.0 to 88.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR