乙酸己酯 ≥99.0% (by GC)

供应商: TCI

Synonyms: 正己基乙酸酯

A0032-25ML A0032-500ML
TCIAA0032-25MLEA 0 CNY
TCIAA0032-25ML TCIAA0032-500ML
乙酸己酯 ≥99.0% (by GC)
乙酸己酯
分子式: C₈H₁₆O₂
沸点: 169°C
溶点: –80 °C
密度: 0,87 g/cm³ (20 °C)
闪点: 62°C
存储条件: Refrigerator
MDL编号: MFCD00009524
CAS号: 142-92-7
EINECS: 205-572-7
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e61aa227bb687c7418ea33167ed1e09fd3b7325&vid=c3d7e3b7722c4f2e6cba91efb93e3a1d74c29e86&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR