乙酸庚酯 ≥99.0% (by GC)
供应商: TCI
Synonyms:
乙酸正庚酯
A0904-25ML
A0904-500ML
TCIAA0904-25MLEA
0
CNY
TCIAA0904-25ML
TCIAA0904-500ML
乙酸庚酯 ≥99.0% (by GC)
乙酸庚酯
分子式:
C₉H₁₈O₂ 分子量: 158.24 g/mol 沸点: 192°C 溶点: –50°C 闪点: 69°C |
MDL编号:
MFCD00027311 CAS号: 112-06-1 EINECS: 203-932-8 |
|
产品技术规格
Appearance | Colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...