5-壬酮 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 二异丁基酮, 二丁基甲酮

N0294-25ML N0294-500ML
TCIAN0294-25MLEA 0 CNY
TCIAN0294-25ML TCIAN0294-500ML
5-壬酮 ≥98.0% (by GC)
5-壬酮
分子式: C₉H₁₈O
沸点: 186°C
溶点: –50 °C
密度: 0,822 g/cm³ (20 °C)
闪点: 60°C
存储条件: Ambient
MDL编号: MFCD00009484
CAS号: 502-56-7
EINECS: 207-946-5
UN号: 1224
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c984c80f53f3ab0bc81da7144a59fee087293af9&vid=bcf8c9c63d239b299a105ba84324614e013e726c&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR