2-壬酮 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
甲基庚基酮
N0293-25ML
N0293-500ML
TCIAN0293-25MLEA
0
CNY
TCIAN0293-25ML
TCIAN0293-500ML
2-壬酮 ≥98.0% (by GC)
2-壬酮
分子式:
C₉H₁₈O 分子量: 142.24 g/mol 沸点: 195°C 溶点: -15 闪点: 76°C 存储条件: Ambient |
MDL编号:
MFCD00009553 CAS号: 821-55-6 EINECS: 212-480-0 |
|
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...