4-Methoxy-2-nitrobenzenesulphonyl chloride ≥93.0% (by titrimetric analysis)

供应商: TCI
Danger

Synonyms: 4-Methoxy-2-nitrobenzene-1-sulphonyl chloride

M1006-25G M1006-5G
TCIAM1006-25GEA 0 CNY
TCIAM1006-25G TCIAM1006-5G
4-Methoxy-2-nitrobenzenesulphonyl chloride ≥93.0% (by titrimetric analysis)
4-Methoxy-2-nitrobenzenesulphonyl chloride
分子式: C₇H₆ClNO₅S
MDL编号: MFCD00143333
CAS号: 18092-54-1
EINECS: 241-996-9
UN号: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=906855d8f8fb9bd93cf8b504d82e2b9e12ab83d5&vid=906855d8f8fb9bd93cf8b504d82e2b9e12ab83d5&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light orange to Yellow to Green powder to crystal
Purity (Argentmetric Titration) min. 93.0 %
Melting point 75.0 to 79.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR