1,4-Diaminobutane dihydrochloride ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 1,4-Butanediamine dihydrochloride, Tetramethylenediammonium dichloride, Tetramethylendiamine dihydrochloride, Putrescine dihydrochloride

D0081-25G D0081-500G
TCIAD0081-25GEA 0 CNY
TCIAD0081-25G TCIAD0081-500G
1,4-Diaminobutane dihydrochloride ≥98.0% (by titrimetric analysis)
1,4-Diaminobutane dihydrochloride
分子式: C₄H₁₄Cl₂N₂
分子量: 161.07 g/mol
溶点: 290°C
MDL编号: MFCD00012526
CAS号: 333-93-7
EINECS: 206-375-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db36cdb25dff162482d60fe6d38c6d334f5bed53&vid=db36cdb25dff162482d60fe6d38c6d334f5bed53&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR