酪胺盐酸盐 ≥98.0% (by titrimetric analysis)
供应商: TCI
|
Warning
|
Synonyms:
4-(2-氨乙基)苯酚, 4-羟基苯乙胺
A0303-1G
A0303-25G
TCIAA0303-1GEA
0
CNY
TCIAA0303-1G
TCIAA0303-25G
酪胺盐酸盐 ≥98.0% (by titrimetric analysis)
酪胺盐酸盐
分子式:
C₈H₁₂ClNO 溶点: 270…272 °C |
MDL编号:
MFCD00012901 CAS号: 60-19-5 EINECS: 200-462-5 Merck Index: 14,09835 |
|
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...