3-苯基-1-丙胺 ≥97.0% (by GC, titration analysis)

供应商: TCI
Danger

Synonyms: γ-氨丙基苯, 3-苯基丙胺

P0664-25G P0664-5G
TCIAP0664-25GEA 0 CNY
TCIAP0664-25G TCIAP0664-5G
3-苯基-1-丙胺 ≥97.0% (by GC, titration analysis)
3-苯基-1-丙胺
分子式: C₉H₁₃N
分子量: 135.21 g/mol
沸点: 220°C
闪点: 90°C
MDL编号: MFCD00008224
CAS号: 2038-57-5
EINECS: 218-012-1
UN号: 1760
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7540b3e8f5caf548c3531a3a1b057f3f9d6eebae&vid=7540b3e8f5caf548c3531a3a1b057f3f9d6eebae&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR