2-(Trifluoromethoxy)benzenesulphonamide ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

T2604-25G T2604-5G
TCIAT2604-25GEA 0 CNY
TCIAT2604-25G TCIAT2604-5G
2-(Trifluoromethoxy)benzenesulphonamide ≥98.0% (by GC, titration analysis)
2-(Trifluoromethoxy)benzenesulphonamide
分子式: C₇H₆F₃NO₃S
分子量: 241.19 g/mol
MDL编号: MFCD01320751
CAS号: 37526-59-3
EINECS: 448-450-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=71d5ef609fa32d328a7b47fc1fc924eb1c8a4cfa&vid=71d5ef609fa32d328a7b47fc1fc924eb1c8a4cfa&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 185.0 to 190.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR