3′,5′-双(三氟甲基)苯乙酮 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 1,3-Di(trifluormethyl)-5-ethanoylbenzol, 1-(3,5-Bis(trifluoromethyl)phenyl)ethanone, 1-[3,5-Bis(trifluoromethyl)phenyl]ethan-1-one

B1949-25G B1949-5G
TCIAB1949-25GEA 0 CNY
TCIAB1949-25G TCIAB1949-5G
3′,5′-双(三氟甲基)苯乙酮 ≥98.0% (by GC)
3′,5′-双(三氟甲基)苯乙酮
分子式: C₁₀H₆F₆O
分子量: 256.15 g/mol
沸点: 95
闪点: 82°C
MDL编号: MFCD00009910
CAS号: 30071-93-3
EINECS: 250-023-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ba8f4fd67a07fd04941b6bd8b0de7256dff4b83&vid=3ba8f4fd67a07fd04941b6bd8b0de7256dff4b83&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR