3-氟-4-甲氧基苯腈 ≥98.0% (by GC)

供应商: TCI
Danger

F0671-1G F0671-5G
TCIAF0671-1GEA 0 CNY
TCIAF0671-1G TCIAF0671-5G
3-氟-4-甲氧基苯腈 ≥98.0% (by GC)
3-氟-4-甲氧基苯腈
分子式: C₈H₆FNO
分子量: 151.14 g/mol
沸点: 98°C
MDL编号: MFCD00215835
CAS号: 331-62-4
UN号: 3439
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6fdc6ca0073ec54275e8c4e8db11a90de5b7be19&vid=6fdc6ca0073ec54275e8c4e8db11a90de5b7be19&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 98.0 to 102.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR