2-Fluoronicotinonitrile ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
3-Cyano-2-fluoropyridine
C2538-25G
C2538-5G
TCIAC2538-25GEA
0
CNY
TCIAC2538-25G
TCIAC2538-5G
2-Fluoronicotinonitrile ≥98.0% (by GC)
2-Fluoronicotinonitrile
分子式:
C₆H₃FN₂ 分子量: 122.1 g/mol 沸点: 196 |
MDL编号:
MFCD03095082 CAS号: 3939-13-7 |
|
产品技术规格
Appearance | White to Dark green powder to lump |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...