5-氟-2-甲氧基苯甲醛 ≥97.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
5-氟邻茴香醛
F0428-1G
F0428-5G
TCIAF0428-1GEA
0
CNY
TCIAF0428-1G
TCIAF0428-5G
5-氟-2-甲氧基苯甲醛 ≥97.0% (by GC)
5-氟-2-甲氧基苯甲醛
分子式:
C₈H₇FO₂ 分子量: 154.14 g/mol 沸点: 118°C 存储条件: Refrigerator |
MDL编号:
MFCD00143458 CAS号: 19415-51-1 |
|
产品技术规格
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 97.0 % |
Melting point | 46.0 to 50.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...