乙酸钯(II) ≥98.0% (通过滴定分析)

供应商: TCI
Danger

A1424-1G A1424-5G
TCIAA1424-1GEA 0 CNY
TCIAA1424-1G TCIAA1424-5G
乙酸钯(II) ≥98.0% (通过滴定分析)
乙酸钯(II)
分子式: C₄H₆O₄Pd
溶点: 205 °C
MDL编号: MFCD00012453
CAS号: 3375-31-3
EINECS: 222-164-4
Merck Index: 14,06991
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e1ff61fdcb945acd33cd2fb856fdbd08f9f6326&vid=2e1ff61fdcb945acd33cd2fb856fdbd08f9f6326&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (Chelometric Titration) 98.0 to 102.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR