(4-Nitrophenyl)acetic acid ethyl ester ≥98.0% (by GC)
供应商: TCI
Synonyms:
Ethyl 4-nitrohenylacetate, Ethyl (4-nitrophenyl)acetate, Ethyl 4-nitrophenylacetate
N0228-25G
N0228-5G
TCIAN0228-25GEA
0
CNY
TCIAN0228-25G
TCIAN0228-5G
(4-Nitrophenyl)acetic acid ethyl ester ≥98.0% (by GC)
(4-Nitrophenyl)acetic acid ethyl ester
分子式:
C₁₀H₁₁NO₄ 分子量: 209.2 g/mol 沸点: 196°C |
MDL编号:
MFCD00017046 CAS号: 5445-26-1 EINECS: 226-646-5 |
|
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 62.0 to 66.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...