4-硝基苯甲酰胺 ≥98.0% (by total nitrogen basis)

供应商: TCI
Warning

N0323-25G N0323-500G
TCIAN0323-25GEA 0 CNY
TCIAN0323-25G TCIAN0323-500G
4-硝基苯甲酰胺 ≥98.0% (by total nitrogen basis)
4-硝基苯甲酰胺
分子式: C₇H₆N₂O₃
分子量: 166.14 g/mol
MDL编号: MFCD00007994
CAS号: 619-80-7
EINECS: 210-613-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4855ad8d09207894c2b53a180f76c61c7d4d99ee&vid=4855ad8d09207894c2b53a180f76c61c7d4d99ee&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 200.0 to 204.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR