3′,5′-二甲氧基-4′-羟基苯乙酮 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
4′-羟基-3′,5′-二甲氧基苯乙酮, 乙酰丁香酮
D2666-25G
D2666-5G
TCIAD2666-25GEA
0
CNY
TCIAD2666-25G
TCIAD2666-5G
3′,5′-二甲氧基-4′-羟基苯乙酮 ≥98.0% (by GC)
3′,5′-二甲氧基-4′-羟基苯乙酮
分子式:
C₁₀H₁₂O₄ 分子量: 196.2 沸点: 335 |
MDL编号:
MFCD00008748 CAS号: 2478-38-8 EINECS: 219-610-5 |
|
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 124.0 to 127.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...